Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UXT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110562
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110562 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110562 Supplier Invitrogen™ Supplier No. PA5110562
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MATPPKRRAV EATGEKVLRY ETFISDVLQR DLRKVLDHRD KVYEQLAKYL QLRNVIERLQ EAKHSELYMQ VDLGCNFFVD TVVPDTSRIY VALGYGFFLE LTLAEALKFI DRKSSLLTEL SNSLTKDSMN IKAHIHMLLE GLRELQGLQN FPEKPHH.

This gene encodes a novel protein which is highly conserved in mouse. It interacts with the N-terminus of the androgen receptor and plays a role in facilitating receptor-induced transcriptional activation. It is also likely to be involved in tumorigenesis as it is abundantly expressed in tumor tissues. This gene is part of a gene cluster on chromosome Xp11.23. Alternative splicing results in 2 transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UXT
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.94 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene UXT
Gene Accession No. Q9UBK9
Gene Alias 0910002B17Rik; androgen receptor trapped clone 27 protein; ART-27; HSPC024; Protein UXT; RP1-212G6.2; SKP2-associated alpha PFD 1; STAP1; ubiquitously expressed prefoldin like chaperone; ubiquitously expressed transcript; ubiquitously expressed transcript protein; ubiquitously-expressed transcript; ubiquitously-expressed, prefoldin-like chaperone; Unknown (protein for MGC:133475); UXT
Gene Symbols UXT
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 13-169 of human UXT (NP_705582.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8409
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.