Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vδ 6.3/2 TCR Hamster anti-Mouse, BUV395, Clone: 8F4H7B7, BD Optibuild™

Catalog No. BDB744478 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744478 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744478 Supplier BD Biosciences Supplier No. 744478
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

The 8F4H7B7 antibody reacts with Vδ6.3/2 T-cell Receptor (TCR)-bearing T lymphocytes. Originally defined as a member of the Vδ6 TCR subfamily, it is now proposed that the C57BL-derived Vδ6.3 is an allelic variant of Vδ6.2, found in A/J, AKR, BALB/c, C3H/He, and FVB mice. mAb 8F4H7B7 crossreacts with Vδ6.4 and possibly Vδ6.6 in DBA/2 mice, and it also detects a subset ofγδTCR-bearing cells in CBA/J and C57L mice. It does not recognize Vδ4, Vδ5, Vδ6.1, or Vδ6.5 TCR. A subpopulation of thymocytes expressing Vδ6.3 or Vδ6.4 TCR (in C57BL/6 or DBA/2 mice, respectively) and low levels of CD90.2 (Thy-1.2) shares functional and phenotypic characteristics with NK-T cells. SimilarδV 6.4 TCRexpressing lymphocytes make up significant proportions of theγδT-cell populations in the liver and spleen of DBA/2 mice. Furthermore, T lymphocytes bearing Vδ6.3/2 TCR are found in the skin and intestinal epithelium and may represent a unique T-cell subpopulation with a potential for autoimmune reactivity.

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vδ 6.3/2 TCR
Classification Monoclonal
Clone 8F4H7B7
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 395
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias V delta 6.3/2 TCR, TCR Vδ 6.3/2, TCR Vδ6.3/2, TCR Vd6.3/2
Host Species Armenian Hamster
Immunogen 1E6.11.D7, a C57BL/6 mouse Vγ1.1Vδ6.3 TCR-bearing IEL/T-cell hybridoma
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.