Learn More
Vβ 8.1, 8.2 TCR Mouse anti-Mouse, Brilliant Violet 605, Clone: MR5-2, BD Optibuild™
Shop All BD Biosciences ProductsDescription
The MR5-2 antibody reacts with the V β 8.1 and V β 8.2 T-cell Receptors (TCR), but not the V β 8.3 TCR, of mice having the b haplotype ( e.g ., A, AKR, BALB/c, CBA/Ca, CBA/J, C3H/He, C57BL, C58, DBA/1, DBA/2) of the Tcrb gene complex. The Tcrb-V8 subfamily gene loci are deleted in mice having the a ( e.g ., C57BR, C57L, SJL, SWR) or c ( e.g., RIII) haplotype. V β 8.1 TCR-bearing T lymphocytes are clonally eliminated in mice expressing superantigen encoded by the Mtv-7 (Mls-1a, Mlsa), provirus ( e.g., AKR, CBA/J, C58, DBA/2), and activation or elimination of V β 8.1 TCR-expressing T cells by this determinant is partially dependent upon presentation by I-E. Mtv-43 (e.g., MA/MyJ), Mtv-44 (e.g., NZW), and/or exogenous MMTV-SW superantigens also cause incomplete elimination of V β 8.1 TCR-bearing T cells. In addition to expression on conventional T lymphocytes, V β 8.2 is the predominant β chain of the TCR on NK-T cells. Staphylococcal enterotoxin B, in association with antigen presenting cells expressing I-A and/or I-E, stimulates lymphocytes bearing V β 8 TCR and selectively eliminates those T cells in vivo . Plate-bound MR5-2 antibody activates V β 8.1 or 8.2 TCR-bearing T lymphocytes.
Specifications
Specifications
| Antigen | Vβ 8.1, 8.2 TCR |
| Classification | Monoclonal |
| Clone | MR5-2 |
| Concentration | 0.2mg/mL |
| Conjugate | Brilliant Violet 605 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | TCR V beta 8.1, TCR V beta 8.2 |
| Host Species | Mouse |
| Immunogen | C57BL/6 mouse helper T-cell clone OI6 |
| Purification Method | Affinity Purified |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.