Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vβ 8 T-Cell Receptor Mouse anti-Mouse, Brilliant Violet 786, Clone: F23.1, BD Optibuild™

Catalog No. BDB742381 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742381 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742381 Supplier BD Biosciences Supplier No. 742381
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The F23.1 antibody specifically reacts with the V β 8.1, V β 8.2, and V β 8.3 T-cell receptors (TCR) of mice having the b haplotype ( e.g., A, AKR, BALB/c, CBA/Ca, CBA/J, C3H/He, C57BL, C58, DBA/1, DBA/2) of the Tcrb gene complex. The Tcrb-V8 subfamily gene loci are deleted in mice having the a ( e.g., C57BR, C57L, SJL, SWR) or c ( e.g., RIII) haplotype. V β 8.1 TCR-bearing T lymphocytes are clonally eliminated in mice expressing superantigen coded by Mtv-7 (Mls-1a, Mlsa) provirus ( e.g., AKR, CBA/J, C58, DBA/2), and activation or elimination of V β 8.1 TCR-expressing T cells by this determinant is partially dependent upon presentation by I-E. Mtv-43 and/or exogenous MMTV-SW superantigens also cause incomplete elimination of V β 8.1 TCR-bearing T cells. In addition to expression on conventional T lymphocytes, V β 8.2 is the predominant β chain of the TCR on NK-T cells. Staphylococcal enterotoxin B, in association with antigen-presenting cells expressing I-A and/or I-E, stimulates lymphocytes bearing V β 8 TCR and selectively eliminates those T cells in vivo . Soluble and plate-bound F23.1 antibody activates V β 8 TCR-bearing T cells, soluble antibody blocks cytolysis mediated by V β 8 TCR-bearing cytotoxic T lymphocytes, and in vivo treatment of neonatal mice can arrest intrathymic maturation of V β 8 TCR-bearing T cells.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vβ 8 T-Cell Receptor
Classification Monoclonal
Clone F23.1
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias TCR V beta 8, TCR V beta 8.1/8.2/8.3
Host Species Mouse
Immunogen BALB.B Mouse Lymph-Node and Spleen T Cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.