Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vγ9 TCR Mouse anti-Human, BB700, Clone: B3, BD OptiBuild™

Catalog No. BDB746204 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB746204 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB746204 Supplier BD Biosciences Supplier No. 746204
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Vγ9 TCR Monoclonal antibody specifically detects Vγ9 TCR in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vγ9 TCR
Applications Flow Cytometry
Classification Monoclonal
Clone B3
Concentration 0.2 mg/mL
Conjugate Brilliant Blue 700 (BB700)
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate (2 to 8°C)
Form Purified
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.