Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
V alpha 24 J alpha 18 TCR Antibody (6B11), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP200267APCCY7
Description
V alpha 24 J alpha 18 TCR Monoclonal specifically detects V alpha 24 J alpha 18 TCR in Human samples. It is validated for Flow Cytometry, Flow (Cell Surface).Specifications
V alpha 24 J alpha 18 TCR | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
Mouse | |
Protein G purified | |
Primary | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
6B11 | |
Flow Cytometry, Flow (Cell Surface) | |
iNKT, iNKT cell, invariant natural killer T cell, invariant NK T cell, invariant NKT cell | |
Peptide epitope in the CDR3 region of V alpha 24 | |
0.1 mL | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction