Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | FGF-8 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
FGF-8 Polyclonal specifically detects FGF-8 in Human, Mouse samples. It is validated for Western Blot.Specifications
| FGF-8 | |
| Polyclonal | |
| Rabbit | |
| Angiogenesis, Cell Cycle and Replication | |
| AIGFKAL6, Androgen-induced growth factor, FGF-8, fibroblast growth factor 8, fibroblast growth factor 8 (androgen-induced), HBGF-8, Heparin-binding growth factor 8, MGC149376 | |
| FGF8 | |
| IgG | |
| 24 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| NP_149354 | |
| 2253 | |
| The immunogen for this antibody is FGF8 - middle region. Peptide sequence LIVETDTFGSRVRVRGAETGLYICMNKKGKLIAKSNGKGKDCVFTEIVLE. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title