Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPL22 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$548.00
Specifications
Antigen | RPL22 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
RPL22 Polyclonal specifically detects RPL22 in Mouse samples. It is validated for Western Blot.Specifications
RPL22 | |
Polyclonal | |
Rabbit | |
P67984 | |
6146 | |
Synthetic peptide corresponding to aa 79-128, from the C-terminal region of mouse RPL22 (NP_033105). The peptide is homologous in human. Peptide sequence SKRYLKYLTKKYLKKNNLRDWLRVVANSKESYELRYFQINQDEEEEEDED. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
EAPHBP15, EBER-associated protein, Epstein-Barr virus small RNA-associated protein, Epstein-Barr-encoded RNA-associated protein, HBP15/L22, heparin-binding protein 15, Heparin-binding protein HBp15, ribosomal protein L22,60S ribosomal protein L22 | |
RPL22 | |
IgG | |
15 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title