Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEC62 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | SEC62 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
SEC62 Polyclonal specifically detects SEC62 in Human samples. It is validated for Western Blot.Specifications
SEC62 | |
Polyclonal | |
Rabbit | |
NP_003253 | |
7095 | |
The immunogen for this antibody is SEC62 - middle region. Peptide sequence KYDKDIKKEKDKGKAESGKEEDKKSKKENIKDEKTKKEKEKKKDGEKEES. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Dtrp1, hTP-1, S.cerevisiae, homolog of, SEC62 homolog (S. cerevisiae), TLOC1, TP-1, translocation protein 1, translocation protein SEC62 | |
SEC62 | |
IgG | |
46 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title