Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTHRC1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | CTHRC1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
CTHRC1 Polyclonal specifically detects CTHRC1 in Mouse samples. It is validated for Western Blot.Specifications
| CTHRC1 | |
| Polyclonal | |
| Rabbit | |
| NP_081054 | |
| 115908 | |
| The immunogen for this antibody is Cthrc1 - C-terminal region. Peptide sequence HRTSSVEGLCEGIGAGLVDVAIWVGTCSDYPKGDASTGWNSVSRIIIEEL. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| collagen triple helix repeat containing 1, collagen triple helix repeat-containing protein 1, Protein NMTC1 | |
| CTHRC1 | |
| IgG | |
| 26 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title