Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$499.50
Specifications
| Antigen | KCP |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
KCP Polyclonal specifically detects KCP in Human samples. It is validated for Western Blot.Specifications
| KCP | |
| Polyclonal | |
| Rabbit | |
| NP_955381 | |
| 375616 | |
| The immunogen for this antibody is KCP - N-terminal region. Peptide sequence AHSSVLAGNSQEQWHPLREWLGRLEAAVMELREQNKDLQTRVRQLESCEC. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| CRIM2, KCP1, kielin/chordin-like protein, NET67 | |
| KCP | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title