Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KRTAP3-3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | KRTAP3-3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19850520
![]() |
Novus Biologicals
NBP19850520UL |
20 μL |
Each for $206.00
|
|
|||||
NBP198505
![]() |
Novus Biologicals
NBP198505 |
100 μL |
Each for $487.50
|
|
|||||
Description
KRTAP3-3 Polyclonal specifically detects KRTAP3-3 in Mouse samples. It is validated for Western Blot.Specifications
KRTAP3-3 | |
Polyclonal | |
Rabbit | |
NP_079800 | |
85293 | |
The immunogen for this antibody is Krtap3-3 - N-terminal region. Peptide sequence SVPTGPATTICSSDKSCRCGVCLPSTCPHTIWQLEPTCCDNCPPPCHIPQ. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
High sulfur keratin-associated protein 3.3, KAP3.3MGC95374, keratin associated protein 3-3, Keratin-associated protein 3.3, keratin-associated protein 3-3, KRTAP3.3 | |
KRTAP3-3 | |
IgG | |
10 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title