Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NPIP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | NPIP |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
NPIP Polyclonal specifically detects NPIP in Human samples. It is validated for Western Blot.Specifications
NPIP | |
Polyclonal | |
Rabbit | |
Neuroscience | |
NP_008916 | |
9284 | |
The immunogen for this antibody is NPIP - middle region. Peptide sequence NRHDKINGKRKTAKEHLRKLSMKEREHGEKERQVSEAEENGKLDMKEIHT. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
FLJ42525, FLJ44848, nuclear pore complex interacting protein, nuclear pore complex-interacting protein | |
NPIP | |
IgG | |
39 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title