Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Artemin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | Artemin |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Artemin Polyclonal specifically detects Artemin in Human samples. It is validated for Western Blot.Specifications
Artemin | |
Polyclonal | |
Rabbit | |
Cancer, Growth and Development, Neuronal Cell Markers, Neuroscience | |
NP_476431 | |
9048 | |
The immunogen for this antibody is Artemin - middle region. Peptide sequence RTARWCSGRARRPPPQPSRPAPPPPAPPSALPRGGRAARAGGPGSRARAA. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
artemin, ENOVIN, EVNneurotrophic factor, NBN, Neublastin | |
ARTN | |
IgG | |
19 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title