Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eukaryotic translation initiation factor 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | Eukaryotic translation initiation factor 1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Eukaryotic translation initiation factor 1 Polyclonal specifically detects Eukaryotic translation initiation factor 1 in Human samples. It is validated for Western Blot.Specifications
Eukaryotic translation initiation factor 1 | |
Polyclonal | |
Rabbit | |
NP_005792 | |
10209 | |
The immunogen for this antibody is Eukaryotic translation initiation factor 1 - middle region. Peptide sequence DKKKLVKAFKKKFACNGTVIEHPEYGEVIQLQGDQRKNICQFLVEIGLAK. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
A121Protein translation factor SUI1 homolog, eIF1, EIF-1, EIF1A, eukaryotic translation initiation factor 1, ISO1, Sui1iso1, SUI1sui1iso1 | |
EIF1 | |
IgG | |
13 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title