Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FKBP10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP198561
Description
FKBP10 Polyclonal specifically detects FKBP10 in Human samples. It is validated for Western Blot.Specifications
FKBP10 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
65 kDa FK506-binding protein, 65 kDa FKBP, EC 5.2.1.8, FK506 binding protein 10 (65 kDa), FK506 binding protein 10, 65 kDa, FK506-binding protein 10, FKBP-10, FKBP6, FKBP-65, FKBP65PPIase FKBP10, FLJ20683, FLJ22041, FLJ23833, hFKBP65, Immunophilin FKBP65, OI6, peptidyl-prolyl cis-trans isomerase FKBP10, PPIASE, Rotamase | |
Rabbit | |
61 kDa | |
100 μL | |
Primary | |
Zebrafish: 86%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_068758 | |
FKBP10 | |
The immunogen for this antibody is FKBP10 - N-terminal region. Peptide sequence VRYHYNGTFEDGKKFDSSYDRNTLVAIVVGVGRLITGMDRGLMGMCVNER. | |
Affinity purified | |
RUO | |
60681 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction