Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRXR3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
| Antigen | TRXR3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19828720
![]() |
Novus Biologicals
NBP19828720UL |
20 μL |
Each for $206.00
|
|
|||||
NBP198287
![]() |
Novus Biologicals
NBP198287 |
100 μL |
Each for $487.50
|
|
|||||
Description
TRXR3 Polyclonal specifically detects TRXR3 in Human samples. It is validated for Western Blot.Specifications
| TRXR3 | |
| Polyclonal | |
| Rabbit | |
| NP_001166984 | |
| 114112 | |
| The immunogen for this antibody is TRXR3 - middle region. Peptide sequence NHISSLNWGYRLSLREKAVAYVNSYGEFVEHHKIKATNKKGQETYYTAAQ. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| TGR, thioredoxin reductase 3, TR2, TRXR3 | |
| TXNRD3 | |
| IgG | |
| 66 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title