Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERP1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP19859220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP19859220 20 μL
NBP198592 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP19859220 Supplier Novus Biologicals Supplier No. NBP19859220UL

Rabbit Polyclonal Antibody

SERP1 Polyclonal specifically detects SERP1 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen SERP1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_055260
Gene Alias FLJ43424, MGC117327, MGC133321, RAMP4MGC133322, ribosome associated membrane protein 4, Ribosome-attached membrane protein 4, stress-associated endoplasmic reticulum protein 1
Gene Symbols SERP1
Host Species Rabbit
Immunogen The immunogen for this antibody is SERP1 - N-terminal region. Peptide sequence IRMANEKHSKNITQRGNVAKTSRNAPEEKASVGPWLLALFIFVVCGSAIF.
Molecular Weight of Antigen 7 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27230
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.