Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Activin C/Inhibin beta C Antibody, Novus Biologicals™
SDP

Catalog No. NBP19829720 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP19829720 20 μL
NBP198297 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP19829720 Supplier Novus Biologicals Supplier No. NBP19829720UL

Rabbit Polyclonal Antibody

Activin C/Inhibin beta C Polyclonal specifically detects Activin C/Inhibin beta C in Human samples. It is validated for Western Blot.

Specifications

Antigen Activin C/Inhibin beta C
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_005529
Gene Alias inhibin beta C chain, IHBC, inhibin, beta C
Gene Symbols INHBC
Host Species Rabbit
Immunogen The immunogen for this antibody is INHBC - N-terminal region. Peptide sequence QECEIISFAETGLSTINQTRLDFHFSSDRTAGDREVQQASLMFFVQLPSN.
Molecular Weight of Antigen 13 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3626
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.