Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAMTOR4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP198263
Description
LAMTOR4 Polyclonal specifically detects LAMTOR4 in Mouse samples. It is validated for Western Blot.Specifications
C7orf59 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
C7orf59, chromosome 7 open reading frame 59, hypothetical protein LOC389541, late endosomal/lysosomal adaptor, MAPK and MTOR activator 4, MGC163425, MGC163431 | |
Rabbit | |
11 kDa | |
100 μL | |
Signal Transduction | |
389541 | |
Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_001074577 | |
LAMTOR4 | |
The immunogen for this antibody is UPF0539 protein C7orf59 N-terminal region. Peptide sequence MTSALTQGLERIPDQLGYLVLSEGAVLASSGDLENDEQAASAISELVSTA. | |
Affinity purified | |
RUO | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction