Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLK4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP19835220UL
Description
CLK4 Polyclonal specifically detects CLK4 in Mouse samples. It is validated for Western Blot.Specifications
CLK4 | |
Polyclonal | |
Western Blot 1:1000 | |
NP_031740 | |
CLK4 | |
The immunogen for this antibody is Clk4 - N-terminal region. Peptide sequence DSDCHYLEARCLNERDYRDRRYIDEYRNDYCEGYVPRHYHRDVESTYRIH. | |
Affinity Purified | |
RUO | |
57396 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS & 2% Sucrose. with No Preservative | |
CDC-like kinase 4EC 2.7.12.1, DKFZp686A20267, dual specificity protein kinase CLK4, protein serine threonine kinase Clk4 | |
Rabbit | |
57 kDa | |
20 μL | |
Primary | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction