Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXB5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$480.74
Specifications
| Antigen | HOXB5 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP198351
![]() |
Novus Biologicals
NBP198351 |
100 μL |
Each for $480.74
|
|
|||||
NBP1983520
![]() |
Novus Biologicals
NBP19835120UL |
20 μL | N/A | N/A | N/A | ||||
Description
HOXB5 Polyclonal specifically detects HOXB5 in Mouse samples. It is validated for Western Blot.Specifications
| HOXB5 | |
| Polyclonal | |
| Rabbit | |
| NP_032294 | |
| 3215 | |
| The immunogen for this antibody is Hoxb5 - middle region. Peptide sequence ATPASSSANFTEIDEASASSEPEEAASQLSSPSLARAQPEPMATSTAAPE. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| HHO.C10, homeo box 2A, homeo box B5, homeobox B5, Homeobox protein HHO.C10, Homeobox protein Hox-2A, homeobox protein Hox-B5, Homeobox protein Hu-1, Hox2.1, HOX2AHOX2, HU-1 | |
| HOXB5 | |
| IgG | |
| 29 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title