Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CP2F2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP198269

 View more versions of this product

Catalog No. NBP198269


Only null left
Add to Cart

Description

Description

CP2F2 Polyclonal specifically detects CP2F2 in Rat samples. It is validated for Western Blot.
Specifications

Specifications

CP2F2
Polyclonal
Unconjugated
PBS, 2% Sucrose with 0.09% Sodium Azide
Cyp2f2
Rabbit
54 kDa
100 μL
Primary
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
NP_062176
Cyp2f2
The immunogen for this antibody is CP2F2 - middle region. Peptide sequence EHQDSLDPNSPRDFIDCFLTKMVQEKQDPLSHFNMDTLLMTTHNLLFGGT.
Affinity purified
RUO
Porcine: 86%; Goat: 86%; Equine: 86%; Mouse: 86%; Bovine: 86%; Canine: 77%.
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Goat, Rabbit
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.