Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
V alpha 24 J alpha 18 TCR Antibody (6B11), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP200267H
Description
V alpha 24 J alpha 18 TCR Monoclonal specifically detects V alpha 24 J alpha 18 TCR in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
V alpha 24 J alpha 18 TCR | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
Mouse | |
Protein G purified | |
Primary | |
Store at 4C in the dark. | |
IgG1 κ |
Immunohistochemistry, Immunohistochemistry (Frozen) | |
6B11 | |
Immunohistochemistry, Immunohistochemistry-Frozen | |
iNKT, iNKT cell, invariant natural killer T cell, invariant NK T cell, invariant NKT cell | |
Peptide epitope in the CDR3 region of V alpha 24 | |
0.1 mL | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction