Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
V alpha 24 J alpha 18 TCR Antibody (6B11), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP200267PCP
Description
V alpha 24 J alpha 18 TCR Monoclonal specifically detects V alpha 24 J alpha 18 TCR in Human samples. It is validated for Flow Cytometry, Flow (Cell Surface).Specifications
V alpha 24 J alpha 18 TCR | |
Monoclonal | |
PerCP | |
iNKT, iNKT cell, invariant natural killer T cell, invariant NK T cell, invariant NKT cell | |
Peptide epitope in the CDR3 region of V alpha 24 | |
0.1 mL | |
Primary | |
Store at 4C in the dark. |
Flow Cytometry | |
6B11 | |
Flow Cytometry, Flow (Cell Surface) | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction