Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VMA2 Monoclonal Antibody (13D11B2)
GREENER_CHOICE

Catalog No. A6427
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A6427 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A6427 Supplier Invitrogen™ Supplier No. A6427
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This clone recognizes the 60kDa subunit 1 (B-subunit, the product of the VMA2 or VAT2 gene) of S. cerevisiae. By Western blot, this can be used to identify organelles during subcellular fractionation of yeast membranes.

Vacuolar-type H+-ATPase (V-ATPase) is a multisubunit enzyme responsible for acidification of eukaryotic intracellular organelles. V-ATPases pump protons against an electrochemical gradient, while F-ATPases reverse the process, thereby synthesizing ATP. A peripheral V1 domain, which is responsible for ATP hydrolysis, and a integral V0 domain, which is responsible for proton translocation, compose V-ATPase. Nine subunits (A-H) make up the V1 domain and five subunits (a, d, c, c' and c ) make up the V0 domain. Like F-ATPase, V-ATPase most likely operates through a rotary mechanism. The V-ATPase V1 B subunit exists as two isoforms. In the inner ear, the V-ATPase B1 isoform functions in proton secretion and is required to maintain proper endolymph pH and normal auditory function. The gene encoding the human V-ATPase B1 isoform maps to chromosome 2cen-q13. Mutations in this gene cause distal renal tubular acidosis associated with sensorineural deafness. The V-ATPase B2 isoform is expressed in kidney and is the only B isoform expressed in osteoclasts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VMA2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 13D11B2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation HEPES buffered saline with 0.02% sodium azide
Gene VMA2
Gene Alias ATP6B1; ATP6V1B1; ATPase H+ transporting V1 subunit B1; ATPase, H transporting, lysosomal V1 subunit B1; ATPase, H+ transporting, lysosomal (vacuolar proton pump), beta 56/58 kDa, isoform 1; ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B, isoform 1; ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B1; ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B1 (Renal tubular acidosis with deafness); ATPase, H+ transporting, lysosomal V1 subunit B1; ATPase, H+ transporting, V1 subunit B, isoform 1; AW208839; D630003L15; D630030L16Rik; D630039P21Rik; endomembrane proton pump 58 kDa subunit; H(+)-transporting two-sector ATPase, 58kD subunit; H+-ATPase beta 1 subunit; lysosomal 56/58kDa; RTA1B; vacuolar H+-ATPase; vacuolar proton pump 3; vacuolar proton pump subunit B 1; vacuolar proton pump, subunit 3; VATB; V-ATPase B1; V-ATPase B1 subunit; V-ATPase subunit B 1; VMA2; VPP3; Vpp-3; V-type proton ATPase subunit B, kidney isoform
Gene Symbols VMA2
Host Species Mouse
Immunogen Purified S. cerevisiae vacuole membranes.
Purification Method Purified
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 852424
Target Species Yeast
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.