Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ V-ATPase C1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566075
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566075 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566075 Supplier Invitrogen™ Supplier No. PA566075
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VVVPKLNHNDW IKQYETLAEM VVPRSSNVLS EDQDSYLCNV TLFRKAVDDF RHKARENKFI VRDFQYNEEE MKADKEEMNR Highest antigen sequence identity to the following orthologs - mouse 99%, rat 98%.

ATP6V1C1 is a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of intracellular compartments of eukaryotic cells. V-ATPase dependent acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c', and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This gene is one of two genes that encode the V1 domain C subunit proteins and is found ubiquitously. This C subunit is analogous but not homologous to gamma subunit of F-ATPases. Previously, this gene was designated ATP6D.
TRUSTED_SUSTAINABILITY

Specifications

Antigen V-ATPase C1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATP6V1C1
Gene Accession No. P21283
Gene Alias 1700025B18Rik; ATP6C; Atp6c1; ATP6D; Atp6v1c1; ATPase H+ transporting V1 subunit C1; ATPase, H+ transporting, lysosomal 42kDa, V1 subunit C1; ATPase, H+ transporting, lysosomal V1 subunit C1; ATPase, H+ transporting, V1 subunit C, isoform 1; FLJ20057; H(+)-transporting two-sector ATPase, subunit C; H+ -ATPase C subunit; H+-transporting ATPase chain C, vacuolar; subunit C of vacuolar proton-ATPase V1 domain; testicular tissue protein Li 223; U13839; vacuolar ATP synthase subunit C; vacuolar H+ -ATPase C subunit; vacuolar proton pump C subunit; vacuolar proton pump subunit C 1; vacuolar proton pump, 42-kD subunit; vacuolar proton-ATPase, subunit C, VI domain; vatC; V-ATPase C subunit; V-ATPase subunit C 1; Vma5; V-type proton ATPase subunit C 1
Gene Symbols ATP6V1C1
Host Species Rabbit
Immunogen Recombinant Human V-ATPase C1. Recombinant protein control fragment (Product #RP-105065).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 528
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.