Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ V-ATPase G1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565342
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565342 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565342 Supplier Invitrogen™ Supplier No. PA565342
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AEIEQYRLQRE KEFKAKEAAA LGSRGSCSTE VEKETQEKMT ILQTYFRQNR DEVLDNLLAF VCDIRPEIHE NYR Highest antigen sequence identity to the following orthologs - mouse 92%, rat 95%.

This protein is a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen V-ATPase G1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATP6V1G1
Gene Accession No. O75348
Gene Alias 1810024D14Rik; AA960677; ATP6G; ATP6G1; ATP6GL; ATP6J; ATP6V1G1; ATPase H+ transporting V1 subunit G1; ATPase, H transporting, lysosomal V1 subunit G1; ATPase, H+ transporting, lysosomal (vacuolar proton pump); ATPase, H+ transporting, lysosomal (vacuolar proton pump), member J; ATPase, H+ transporting, lysosomal 13kD, V1 subunit G; ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G1; ATPase, H+ transporting, lysosomal V1 subunit G1; ATPase, H+ transporting, V1 subunit G; DKFZp547P234; lysosomal 13kDa; vacuolar ATP synthase subunit M16; vacuolar H(+)-ATPase subunit G 1; vacuolar proton pump subunit G 1; Vacuolar proton pump subunit M16; VAG1; V-ATPase 13 kDa subunit 1; V-ATPase subunit G 1; Vma10; V-type proton ATPase subunit G 1
Gene Symbols ATP6V1G1
Host Species Rabbit
Immunogen Recombinant Human V-ATPase G1. Recombinant protein control fragment (Product #RP-104763).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9550
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.