Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ V-ATPase G1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112349
Description
V-ATPase G1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
This protein is a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation.
Specifications
| V-ATPase G1 | |
| Polyclonal | |
| Unconjugated | |
| ATP6V1G1 | |
| 1810024D14Rik; AA960677; ATP6G; ATP6G1; ATP6GL; ATP6J; ATP6V1G1; ATPase H+ transporting V1 subunit G1; ATPase, H transporting, lysosomal V1 subunit G1; ATPase, H+ transporting, lysosomal (vacuolar proton pump); ATPase, H+ transporting, lysosomal (vacuolar proton pump), member J; ATPase, H+ transporting, lysosomal 13kD, V1 subunit G; ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G1; ATPase, H+ transporting, lysosomal V1 subunit G1; ATPase, H+ transporting, V1 subunit G; DKFZp547P234; lysosomal 13kDa; vacuolar ATP synthase subunit M16; vacuolar H(+)-ATPase subunit G 1; vacuolar proton pump subunit G 1; Vacuolar proton pump subunit M16; VAG1; V-ATPase 13 kDa subunit 1; V-ATPase subunit G 1; Vma10; V-type proton ATPase subunit G 1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 9550 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O75348 | |
| ATP6V1G1 | |
| Recombinant Human V-type proton ATPase subunit G 1 protein (2-118AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction