Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ V-ATPase G3 Monoclonal Antibody (3A5)
GREENER_CHOICE

Catalog No. PIMA523000
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523000 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA523000 Supplier Invitrogen™ Supplier No. MA523000
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: EEAMVEIDQY RMQRDKEFRL KQSKIMGSQN NLSDEIEEQT LGKIQELNGH YNKYMESVMN QLLSMVCDMK PEIHVNYRAT N.

ATP6V1G3 (V-ATPase G3) is a catalytic subunit of the peripheral V1 complex of vacuolar ATPase (V-ATPase). V-ATPase is responsible for acidifying a variety of intracellular compartments in eukaryotic cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen V-ATPase G3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3A5
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ATP6V1G3
Gene Accession No. Q96LB4
Gene Alias ATP6G3; ATP6V1G3; ATPase H+ transporting V1 subunit G3; ATPase, H+ transporting, lysosomal (vacuolar proton pump) subunit G3; ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G3; ATPase, H+ transporting, lysosomal V1 subunit G3; ATPase, H+ transporting, lysosomal, V1 subunit G3; ATPase, H+ transporting, V1 subunit G, isoform 3; vacuolar ATP synthase subunit G 3; vacuolar proton pump G subunit 3; vacuolar proton pump subunit G 3; vacuolar proton pump, subunit G3; V-ATPase 13 kDa subunit 3; V-ATPase G subunit 3; V-ATPase G3 subunit; V-ATPase subunit G 3; Vma10; V-type proton ATPase subunit G 3
Gene Symbols ATP6V1G3
Host Species Mouse
Immunogen ATP6V1G3 (NP_573569, 38 a.a. ~ 118 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127124
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.