Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ V-ATPase H Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599738
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599738 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599738 Supplier Invitrogen™ Supplier No. PA599738
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total V-ATPase H.

This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of intracellular organelles. V-ATPase-dependent organelle acidification is necessary for multiple processes including protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. The encoded protein is the regulatory H subunit of the V1 domain of V-ATPase, which is required for catalysis of ATP but not the assembly of V-ATPase. Decreased expression of this gene may play a role in the development of type 2 diabetes. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen V-ATPase H
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ATP6V1H
Gene Accession No. Q8BVE3, Q9UI12
Gene Alias 0710001F19Rik; Atp6v1h; ATPase H+ transporting V1 subunit H; ATPase, H+ transporting, lysosomal 50/57kDa, V1 subunit H; ATPase, H+ transporting, lysosomal V1 subunit H; AU022349; CGI-11; MSTP042; NBP1; Nef-binding protein 1; protein VMA13 homolog; SFD; SFDalpha; SFDbeta; vacuolar ATP synthase subunit H; vacuolar ATPase subunit H; vacuolar proton pump H subunit; vacuolar proton pump subunit H; Vacuolar proton pump subunit SFD; V-ATPase 50/57 kDa subunits; V-ATPase H subunit; V-ATPase subunit H; VMA13; V-type proton ATPase subunit H
Gene Symbols ATP6V1H
Host Species Rabbit
Immunogen A synthesized peptide derived from human ATP6V1H(Accession Q9UI12), corresponding to amino acid residues F337-N387.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108664, 51606
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.