Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V-ATPase Mouse anti-Yeast, Clone: 13D11B2, Abnova™

Catalog No. 89035586 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-586 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-586 Supplier Abnova Corporation Supplier No. MAB14189

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against yeast V-ATPase.

  • High specificity
  • High reactivity

Specifications

Antigen V-ATPase
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 13D11B2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against yeast V-ATPase.
Dilution Immunofluorescence Western Blot The optimal working dilution should be determined by the end user.
Formulation In HEPES (0.02% sodium azide).
Host Species Mouse
Immunogen Yeast V-ATPase.
Purification Method Purified
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Yeast
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.