Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V Bta 11 TCR Mouse, anti-Mouse, R718, Clone: RR3-15, BD OptiBuild™

Catalog No. BDB752015 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752015 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752015 Supplier BD Biosciences Supplier No. 752015
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The RR3-15 antibody reacts with the Vβ 11 T-Cell Receptor (TCR) of mice having the b haplotype (e.g., A, C57BL, C58, DBA/1) of the Tcrb gene complex. The Tcrb-V11 gene locus is deleted in mice having the a (e.g., C57BR, C57L, SJL, SWR) and c (e.g., RIII) haplotypes. Vβ TCR-bearing T lymphocytes are clonally eliminated in mice expressing I-E and superantigens encoded by Mtv-9 (Etc-1, Mls[f], Dvb11.2) and/or Mtv-11 (Mls[f], Dvb 11.2) proviruses (e.g., AKR, BALB/c, CBA/J, C3H, DBA/2), and they are incompletely eliminated in mice expressing I-E and Mtv-8 (Mls[f], Dvb 11.1) superantigen (e.g., A). Activation of Vβ 11 TCR-expressing T cells by these determinants is dependent upon presentation by I-E. The bacterial superantigen Staphylococcal enterotoxin A (SEA) also interacts with Vβ 11 TCR, and in vivo exposure to SEA causes activation and subsequent deletion of Vβ TCR-expressing lymphocytes. Plate-bound RR3-15 antibody activates Vβ 11 TCR-bearing T cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen V Bta 11 TCR
Applications Flow Cytometry
Classification Monoclonal
Clone RR3-15
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.