Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V Bta 12 TCR Mouse, anti-Mouse, R718, Clone: MR11-1, BD OptiBuild™

Catalog No. BDB751958 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751958 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751958 Supplier BD Biosciences Supplier No. 751958
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MR11-1 monoclonal antibody specifically recognizes the Vβ 12 T-cell Receptor (TCR) of mice having the b haplotype (e.g., C57BL, C58, DBA/1) of the Tcrb gene complex1 The Tcrb-V12 gene locus is deleted in mice having the a (e.g., C57BR, C57L, SJL, SWR) or c (e.g., RIII) haplotype. Vβ 12 TCR-bearing T lymphocytes are clonally eliminated in mice expressing I-E and superantigens encoded by Mtv-8 (Mlsf, Dvb11.1), Mtv-9 (Etc-1, Mlsf, Dvb11.2) and/or Mtv-11 (Mlsf, Dvb11.3) proviruses (eg, A, AKR, BALB/c, CBA/J, C3H, DBA/2). Activation of Vβ 12 TCR-expressing T cells by these determinants is dependent upon presentation by I-E. C57BL/6 spleen T cells expressing Vβ 12 TCR are among the predominant responders to MAIDS virus superantigen.

TRUSTED_SUSTAINABILITY

Specifications

Antigen V Bta 12 TCR
Applications Flow Cytometry
Classification Monoclonal
Clone MR11-1
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.