Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V Bta 4 TCR Mouse, anti-Mouse, R718, Clone: KT4, BD OptiBuild™

Catalog No. BDB751911 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751911 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751911 Supplier BD Biosciences Supplier No. 751911
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The KT4 antibody specifically recognizes the Vβ4 T-cell Receptor (TCR) of mice having the a (e.g., C57BR, C57L, SJL, SWR), b (e.g., A, AKR, BALB/c, CBA, C3H/He, C57BL, C58, DBA/1, DBA/2), and c (e.g., RIII) haplotypes of the Tcrb gene complex. Vβ4 TCR-bearing cells are among the CD4+ autoreactive T cells which induce autoimmune thyroiditis after elimination of regulatory (CD5-bright) T-cell subsets. Plate-bound KT4 antibody activates Vβ4 TCR-bearing T cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen V Bta 4 TCR
Applications Flow Cytometry
Classification Monoclonal
Clone KT4
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.