Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V Bta 5.1 5.2 TCR Mouse, anti-Mouse, R718, Clone: MR9-4, BD OptiBuild™

Catalog No. BDB751882 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751882 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751882 Supplier BD Biosciences Supplier No. 751882
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MR9-4 monoclonal antibody specifically recognizes the Vβ 5.1 and Vβ 5.2 T-cell Receptors of strains having the b haplotype (e.g., C57BL) of the Tcrb gene complex. These gene loci are deleted in mice having the a (e.g., C57BR, C57L, SJL, SWR) or c (e.g., RIII) Tcrb haplotype. Vβ5.1 and 5.2 TCR-bearing T lymphocytes are clonally eliminated, either completely or partially, in mice expressing I-E and superantigens encoded by the Mtv-1 (Mls-4a, Mlsc), Mtv-3 (Mlsc), Mtv-8 (Mlsf), Mtv-9 (Etc-1, Mlsf), Mtv-11 (Mlsf), Mtv-13 (Mls-2a, Mlsc), Mtv-27, Mtv44 , and/or Mtv-MAI endogenous provirus (e.g., A, AKR, BALB/c, C3H/He, C58, CBA/Ca, CBA/J, DBA/2, NZB, NZW). Activation of Vβ5 TCR-expressing T cells by this determinant is dependent upon presentation by I-E. Plate-bound MR9-4 antibody activates Vβ5.1 or 5.2 TCR-bearing T cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen V Bta 5.1 5.2 TCR
Applications Flow Cytometry
Classification Monoclonal
Clone MR9-4
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.