Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V Bta 9 TCR Mouse, anti-Mouse, R718, Clone: MR10-2, BD OptiBuild™

Catalog No. BDB751987 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751987 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751987 Supplier BD Biosciences Supplier No. 751987
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MR10-2 monoclonal antibody reacts with the Vβ9 T-cell Receptor (TCR) of mice having the b haplotype (e.g., A, BALB/c, CBA/Ca, C3H/He, C57BL) of the Tcrb gene complex. The Tcrb-V9 gene locus is deleted in mice having the a (e.g., C57BR, C57L, SJL, SWR) or c (e.g., RIII) haplotype. Vβ9-bearing T lymphocytes are clonally eliminated in mice expressing superantigen encoded by Mtv-7 (Mls-1a, Mlsa) provirus (e.g., AKR, CBA/J, DBA/2), and activation or elimination of Vβ9 TCR-expressing T cells by this determinant is partially dependent upon presentation by I-E. Mtv-43 (e.g., MA/MyJ), Mtv-44 (e.g., NZW), and/or exogenous MMTV-SW superantigens also cause incomplete elimination of Vβ9 TCR-bearing T cells. Plate-bound MR10-2 antibody activates Vβ9 TCR-bearing T cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen V Bta 9 TCR
Applications Flow Cytometry
Classification Monoclonal
Clone MR10-2
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.