Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V Dta 4 TCR Mouse, anti-Mouse, R718, Clone: GL2, BD OptiBuild™

Catalog No. BDB752210 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752210 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752210 Supplier BD Biosciences Supplier No. 752210
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The GL2 monoclonal antibody specifically reacts with Vδ 4 T-cell Receptor (TCR)-bearing T cells, which are the predominant γδ TCR-bearing cells in the adult intestinal epithelium (γδ IEL). The frequency of Vδ 4 TCR-bearing γδ IEL differs among inbred strains of mice and may be influenced by haplotypes of the MHC, Tcrg, and/or Tcrd gene complexes. There is evidence that these γδ IEL develop in the absence of thymic influence. T lymphocytes expressing Vδ 4 TCR have also been found in lactating mammary glands, the spleen, and the thymus.

TRUSTED_SUSTAINABILITY

Specifications

Antigen V Dta 4 TCR
Applications Flow Cytometry
Classification Monoclonal
Clone GL2
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.