Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ V-FLEX™ Safety Harness

Catalog No. 19001529 Shop All MSA Products
Change view
Click to view available options
Buckle Style (Leg):
Quick-Connect
Tongue
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Buckle Style (Leg)
19-001-529 Quick-Connect
19-001-530 Tongue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-001-529 Supplier MSA™ Supplier No. 10196069

The V-FLEX™ full body harness is a premium member of the V-Series family of fall protection

  • Exclusive design reduces need for harness adjustment throughout the day, allowing for more comfort and freedom of movement
  • Contoured padding adjusts to shape of your shoulder to minimize fatigue
  • Fewer stitch points saves time during pre-use inspection process
  • Thumb loops on shoulder strap allows for easy harness adjustment and fit
  • Integrated Safety Steps are stowed away in leg pads for suspension trauma prevention in a fall event
  • RaceFLEX™ connection makes donning your harness simple - like buckling a seatbelt

Specifications

Certifications/Compliance ANSI Z359.11-2014 CSA Z259.10
Includes Harness
Size Medium/Large
For Use With (Application) Working at Heights
Product Type Fall Protection
D-Ring Location Back
Capacity (Metric) 181 kg
Buckle Style (Leg) Quick-Connect
Webbing Material Polyester
Buckle Style (Chest) RaceFLEX™ Quick-Connect Buckle
Weight (English) 3.75 lb.
Capacity (English) 400 lb.
Length (Metric) 30.5 cm
Length (English) 12 in.
Style Vest with shoulder padding
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.