Learn More
MSA™ V-Gard™ Protective Slotted Cap with Staz-On™ Suspension
Lightweight for all-day wearing comfort
$9.57 - $17.04
Specifications
Certifications/Compliance Meets or exceeds ANSI Z89.1-2009; Type I; Class E; SEI certified.
Suspension Type Staz-On Pin-Lock
View More Specs
Products 10
Catalog Number Mfr. No. Color Size Price Quantity & Availability  
Catalog Number Mfr. No. Color Size Price Quantity & Availability  
19-062899
Accessories View Documents
MSA™
463945
Orange Standard
Each for $15.14
Only null left
Add to Cart
19-080144
Accessories View Documents
MSA™
488148
Hi-vis Orange Standard
Each for $10.33
Only null left
Add to Cart
19-058270
Accessories View Documents
MSA™
463946
Green Standard
Each for $15.14
Only null left
Add to Cart
18-999-3987
View Documents
MSA™
488398
Purple Standard
Each for $17.04
Only null left
Add to Cart
18-999-3985
View Documents
MSA™
475235
Black Standard
Each for $17.04
Only null left
Add to Cart
19-815-529
View Documents
MSA™
485364
Pink Standard
Each for $17.04
Only null left
Add to Cart
19-054727
Accessories View Documents
MSA™
463942
White Standard
Each for $9.57
Only null left
Add to Cart
19-054725
Accessories View Documents
MSA™
463943
Blue Standard
Each for $17.04
Only null left
Add to Cart
19-054728
Accessories View Documents
MSA™
463944
Yellow Standard
Each for $17.04
Only null left
Add to Cart
19-054726
Accessories View Documents
MSA™
463947
Red Standard
Each for $17.04
Only null left
Add to Cart
Description
Description
  • Four-point webbed Staz-On suspension with perforated vinyl sweatband
  • Sizes: Small (6-7 1/8), Standard (6 1/2-8), and Large (7-8 1/2)
  • Available in more than 20 colors, with custom imprinting and striping available
†With lamp bracket and cord holder (Class C)
Order Info
Contact your Fisher Safety Customer Safety Representative for ordering information for sizes and colors not shown.
Specifications
Specifications
Meets or exceeds ANSI Z89.1-2009; Type I; Class E; SEI certified.
Staz-On Pin-Lock
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.