Description
- Polyethylene shell provides superior impact protection
- Self-adjusting crown straps ensure a comfortable fit
- Lightweight and well-balanced for all-day wearing comfort
- Slotted design works with MSA accessories
- Standard size (6½ to 8)
Suspension System Options:
- 1-Touch™ Suspension
- One-handed squeeze opening
- One-hand slide adjustment
- Snap closure
- Adjustable dual nylon straps
- Staz-On Suspension
- 4-point suspension system helps keep helmet in place
- Features high-quality sweatband and nylon height adjusters
- Easily clips into V Gard hat or cap shell
- Adjustable liner
- Fas-Trac III Suspension
- Flush rear lug attachments eliminate pressure points
- Three levels of nape strap adjustment for customized fit
- Graspable ratchet with ultra-smooth rotation and steadfast hold
Specifications
Specifications
| Color | Yellow |
| Size | Standard |
| Suspension Type | Fas-Trac ratchet suspension |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.