Learn More
MSA™ V-Gard™ Slotted Full-Brim Hats
Catalog No. 189993885 Shop All MSA Products
Change view
Click to view available options
Suspension Type:
1-Touch suspension
Fas-Trac ratchet suspension
Fas-Trac suspension
Staz-On suspension
Color:
Gray
Green
Hi-viz Yellow/Green
Navy Gray
Red
White
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Suspension Type Color
18-999-3885 Fas-Trac suspension Green
18-999-3956 Staz-On suspension Gray
19-130-3438 1-Touch suspension Hi-viz Yellow/Green
19-130-3446 1-Touch suspension White
18-999-3882 Fas-Trac suspension Red
18-999-3880 Fas-Trac suspension White
18-999-3884 Fas-Trac suspension Navy Gray
18-999-3883 Fas-Trac ratchet suspension Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 18-999-3885 Supplier MSA™ Supplier No. 475370
Add to Cart
Add to Cart
This item is not returnable. View return policy
Full-brim hardhat available in multiple, fade-resistant colors
  • Polyethylene shell provides superior impact protection
  • Self-adjusting crown straps ensure a comfortable fit
  • Lightweight and well-balanced for all-day wearing comfort
  • Slotted design works with MSA accessories
  • Standard size (6½ to 8)
Suspension System Options:
  • 1-Touch™ Suspension
    • One-handed squeeze opening
    • One-hand slide adjustment
    • Snap closure
    • Adjustable dual nylon straps
  • Staz-On Suspension  
    • 4-point suspension system helps keep helmet in place
    • Features high-quality sweatband and nylon height adjusters
    • Easily clips into V Gard hat or cap shell
    • Adjustable liner
  • Fas-Trac III Suspension
    • Flush rear lug attachments eliminate pressure points
    • Three levels of nape strap adjustment for customized fit
    • Graspable ratchet with ultra-smooth rotation and steadfast hold
Specifications
Color Green
Size Standard
Suspension Type Fas-Trac suspension
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.