Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V Gma 3 TCR Mouse, anti-Mouse, R718, Clone: 536, BD OptiBuild™

Catalog No. BDB752097 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752097 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752097 Supplier BD Biosciences Supplier No. 752097
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 536 monoclonal antibody specifically recognizes Vγ 3† T-cell Receptor (TCR)-bearing T lymphocytes, which are the predominant γδ TCR-bearing cells in the early fetal thymus and the adult epidermis of euthymic mice. The first T cells to mature in the embryonic thymus express the Vγ 3 and Vδ 1 TCR chains, and their development is dependent upon IL-7. There is evidence that the Vγ 3 TCR-bearing fetal thymocytes are the precursors of the majority of dendritic epidermal T cells (DEC), which may be replenished in the adult by proliferation in situ rather than by seeding from primary lymphoid organs. Although the Vγ 3 TCR is almost exclusively found in the DEC population, it has been shown that the homing of DEC to the epidermis does not require expression of the Vγ 3 gene segment. Vγ 3 TCR-bearing dermal dendritic cells have also been described. Vγ 3 TCR has also been found on a subset of T lymphocytes in the lactating mammary gland and at the site of antigenic challenge in contact-sensitized mice. Plate-bound 536 antibody activates Vγ 3 TCR-bearing T cells, and Fab fragments of mAb 536 block in vitro stimulation of DEC by keratinocytes. † Please note that the Vγ 3 designation correlates with the nomenclature of Garman, Doherty, and Raulet| the Vγ 5 designation of Heilig and Tonegawa is equivalent.

TRUSTED_SUSTAINABILITY

Specifications

Antigen V Gma 3 TCR
Applications Flow Cytometry
Classification Monoclonal
Clone 536
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.