Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ v-Myb Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552611
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552611 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552611 Supplier Invitrogen™ Supplier No. PA552611
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EANAVLSSLQ TIPEFAETLE LIESDPVAWS DVTSFDISDA AASPIKSTPV KLMRIQHNEG AMECQFNVSL VLEGKKNTCN GGNSEAVPLT SPNIAKFSTP PAILRKKRKM RVGHSPGSEL RDGSLNDGGN Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 85%.

A-Myb is expressed at specific times in reproductive tissues, some neural cells, and a subset of normal and neoplastic B lymphocytes. Both A-Myb and B-Myb are expressed in t(14:18) lymphoma cells where they then inhibit cell arrest and apoptotic signaling. Expression of B-Myb rescues cells from p53-induced G1 phase arrest that is mediated by p21, while A-Myb functions as an anti-apoptotic factor by effectively activating the bcl-2 promoter and thereby up-regulating Bcl-2 expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen v-Myb
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYBL1
Gene Accession No. P10243
Gene Alias AMYB; A-myb; G1-419-6; MYB proto-oncogene like 1; MYBA; MYBL1; myb-like protein 1; Myb-related protein A; myeloblastosis oncogene-like 1; repro9; VMYB; v-myb avian myeloblastosis viral oncogene homolog-like 1
Gene Symbols MYBL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human v-Myb. Recombinant protein control fragment (Product #RP-89420).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4603
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.