Learn More
Pyramex™ V2G™ Readers Safety Eyewear
Available in three magnifying strengths
$174.94 - $177.25
Specifications
Lens Type H2X
Lens Tint Clear
Frame Color Black
Includes Strap
Lens Material Polycarbonate
View More Specs
Products 3
Catalog Number Mfr. No. Magnification Power Price Quantity & Availability  
Catalog Number Mfr. No. Magnification Power Price Quantity & Availability  
19-155-330
View Documents
Pyramex™
GB1810STR15
+1.50
Case of 6 for $177.25
Only null left
Add to Cart
19-155-331
View Documents
Pyramex™
GB1810STR20
+2.00
Case of 6 for $177.25
Only null left
Add to Cart
19-155-332
View Documents
Pyramex™
GB1810STR25
+2.50
Case of 6 for $174.94
Only null left
Add to Cart
Description
Description
  • Flame resistant foam padding
  • Lens and frame vented to increase air flow
  • Interchangeable temples and headband included
  • 9.75 base curved lens provides full side protection
  • Headband and temples feature ratchet adjustment for perfect fit
  • Features quick fit retainer
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Ballistic options pass MIL-PRF 32432 High Velocity Impact Standards
  • Specifications
    Specifications
    H2X
    Black
    Polycarbonate
    Clear
    Strap
    PVC
    Videos
    SDS
    Documents
    Documents
    Product Certifications
    Product Content Correction
    Your input is important to us. Please complete this form to provide feedback related to the content on this product.
    Product Title
    Your feedback has been submitted: Thank you for helping us improve our website.