Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V450 Rabbit Anti-Active Caspase-3, BD Horizon™

Catalog No. BDB570312 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570312 25 μg
BDB570306 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB570312 Supplier BD Biosciences Supplier No. 570312
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Active Caspase-3 Monoclonal antibody specifically detects Active Caspase-3 in Human|Mouse samples. It is validated for Intracellular staining (flow cytometry)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Active Caspase-3
Applications Flow Cytometry
Classification Monoclonal
Clone C92-605.rMAb
Concentration 0.2 mg/mL
Conjugate Violet Fluor 450 (V450)
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Gene Alias Apopain; CASP-3; CASP3; CC3; CPP-32; CPP32; Caspase 3; Yama
Host Species Rabbit
Immunogen Human Active Caspase-3 Fragment
Purification Method Affinity Chromatography
Quantity 25 μg
Regulatory Status RUO
Gene ID (Entrez) 836, 12367
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.