Learn More
V5 Epitope Tag Alexa Fluor 647-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | V5 Epitope Tag |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 1036H |
| Conjugate | Alexa Fluor 647 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. |
| Gene Accession No. | Y138513 |
| Gene Alias | GKPIPNPLLGLDST Epitope Tag, Parainfluenza virus 5 V/P, Paramyxovirus SV5, V5 epitope tag |
| Host Species | Rabbit |
| Show More |
For Research Use Only (RUO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.