Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
V5 Epitope Tag Antibody, PE/Cy5.5, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB600381PECY55
Description
V5 Epitope Tag Polyclonal antibody specifically detects V5 Epitope Tag in Epitope Tag samples. It is validated for Flow Cytometry.Specifications
V5 Epitope Tag | |
Polyclonal | |
Flow Cytometry | |
V5 epitope tag | |
Rabbits were immunized with a synthetic peptide representing amino acid residues 95 to 108 of RNA polymerase alpha subunit of simion virus 5 conjugated to KLH. | |
0.1 mL | |
Store at 4°C in the dark. Do not freeze. | |
Epitope Tag |
Flow Cytometry | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
Rabbit | |
Affinity Purified | |
Primary | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction