Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V5 Epitope Tag Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8926
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8926 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB8926 Supplier R&D Systems Supplier No. MAB8926
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

V5 Epitope Tag Monoclonal specifically detects V5 Epitope Tag in Multi-Species samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunoprecipitation.

Specifications

Antigen V5 Epitope Tag
Applications Western Blot, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone 1036H
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunoprecipitation 1-4 ug/100 ug cell lysate
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with Sodium Azide
Gene Accession No. Y138513
Gene Alias GKPIPNPLLGLDST Epitope Tag, Parainfluenza virus 5 V/P, Paramyxovirus SV5, V5 epitope tag
Host Species Rabbit
Immunogen V5 epitope tag peptide Accession # Y138513
Purification Method Protein A or G purified from cell culture supernatant
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects N-terminal and C-terminal V5 epitope-tagged proteins in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS
Target Species Multi-species
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.