Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
V5 Epitope Tag Antibody (SV5-P-K), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP252703JF549
Description
V5 Epitope Tag Monoclonal antibody specifically detects V5 Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunofluorescence.Specifications
| V5EpitopeTag | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Mouse | |
| Protein A or G purified | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| SV5-P-K | |
| Immunohistochemistry, Immunohistochemistry-Frozen, Immunofluorescence | |
| V5 epitope tag | |
| Pk (V5) Epitope Tag (GKPIPNPLLGLDST). | |
| 0.1 mL | |
| Recognises a 14 amino acid sequence (GKPIPNPLLGLDST) derived from the P and V proteins of the paramyxoVirus simian virus 5. | |
| Purified | |
| Epitope Tag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction